tert-Butyl-4-amino-3-(trifluoromethyl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115460-250MG 115460-500MG 115460-1G
102899-382EA 4120 USD
102899-382 102899-384 102899-386
tert-Butyl-4-amino-3-(trifluoromethyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-amino-3-(trifluoromethyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₉F₃N₂O₂
MW: 268.28 g/mol
MDL Number: MFCD11848147
CAS Number: 1255098-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR