tert-Butyl-3-(aminomethyl)-4-fluoropiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115459-250MG 115459-500MG 115459-1G
102896-334EA 2746.15 USD
102896-334 102896-336 102896-338
tert-Butyl-3-(aminomethyl)-4-fluoropiperidine-1-carboxylate ≥95%
tert-Butyl-3-(aminomethyl)-4-fluoropiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₁FN₂O₂
MW: 232.3 g/mol
MDL Number: MFCD18909852
CAS Number: 1303973-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR