1-Benzyl-3-(trifluoromethyl)piperidin-4-ol ≥95%

Supplier: Matrix Scientific

115457-250MG 115457-500MG 115457-1G
102896-304EA 2196.92 USD
102896-304 102896-306 102896-308
1-Benzyl-3-(trifluoromethyl)piperidin-4-ol ≥95%
1-Benzyl-3-(trifluoromethyl)piperidin-4-ol

Hazard: Irritant

Formula: C₁₃H₁₆F₃NO
MDL Number: MFCD18800733
CAS Number: 373603-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR