(1-Benzyl-4-fluoropiperidin-3-yl)methanol ≥95%

Supplier: Matrix Scientific

115454-250MG 115454-500MG
102896-286EA 2196.92 USD
102896-286 102896-288
(1-Benzyl-4-fluoropiperidin-3-yl)methanol ≥95%
(1-Benzyl-4-fluoropiperidin-3-yl)methanol

Hazard: Irritant

Formula: C₁₃H₁₈FNO
MDL Number: MFCD21602012
CAS Number: 1356338-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR