1-Benzyl-3-(difluoromethylene)piperidine ≥95%

Supplier: Matrix Scientific

115453-250MG 115453-500MG 115453-1G
102896-280EA 2816.92 USD
102896-280 102896-282 102896-284
1-Benzyl-3-(difluoromethylene)piperidine ≥95%
1-Benzyl-3-(difluoromethylene)piperidine

Hazard: Irritant

Formula: C₁₃H₁₅F₂N
MDL Number: MFCD21601950
CAS Number: 1356342-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR