[1,2,3]Triazolo[1,5-a]pyridine ≥95%

Supplier: Matrix Scientific

115451-1G 115451-5G 115451-25G
102896-268EA 344.62 USD
102896-268 102896-270 102896-272
[1,2,3]Triazolo[1,5-a]pyridine ≥95%
[1,2,3]Triazolo[1,5-a]pyridine

Hazard: Irritant

Formula: C₆H₅N₃
MDL Number: MFCD00185587
CAS Number: 274-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR