2,2-Difluorothioacetamide ≥94%

Supplier: Matrix Scientific

115449-500MG 115449-1G 115449-5G
102896-256EA 756.92 USD
102896-256 102896-258 102896-260
2,2-Difluorothioacetamide ≥94%
2,2-Difluorothioacetamide

Hazard: Irritant

Formula: C₂H₃F₂NS
MDL Number: MFCD12026308
CAS Number: 180002-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR