Diethyl-2,2-difluoro-4-methylenepentanedioate ≥97%

Supplier: Matrix Scientific

115442-100MG 115442-250MG 115442-500MG
102899-324EA 769.23 USD
102899-324 102899-326 102899-328
Diethyl-2,2-difluoro-4-methylenepentanedioate ≥97%
Diethyl-2,2-difluoro-4-methylenepentanedioate

Hazard: Irritant

Formula: C₁₀H₁₄F₂O₄
MDL Number: MFCD22199152
CAS Number: 1307857-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR