1,3-Dichloro-6-fluoroisoquinoline ≥97%

Supplier: Matrix Scientific

115441-500MG 115441-1G
102899-320EA 275.38 USD
102899-320 102899-322
1,3-Dichloro-6-fluoroisoquinoline ≥97%
1,3-Dichloro-6-fluoroisoquinoline

Hazard: Irritant

Formula: C₉H₄Cl₂FN
MW: 216.0400 g/mol
MDL Number: MFCD11656163
CAS Number: 1041423-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR