1,1-Bis(trimethylsilyloxy)-1-propene ≥97%
Supplier: Matrix Scientific
115439-500MG
115439-1G
102896-196EA
176.54
USD
102896-196
102896-198
1,1-Bis(trimethylsilyloxy)-1-propene ≥97%
1,1-Bis(trimethylsilyloxy)-1-propene
Hazard: Irritant
Formula:
C₉H₂₂O₂Si₂ |
MDL Number:
MFCD01632569 CAS Number: 31469-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...