Imidazo[1,2-a]pyridine-6-carbaldehyde ≥95%
Supplier: Matrix Scientific
115436-1G
115436-5G
102896-186EA
1238.46
USD
102896-186
102896-188
Imidazo[1,2-a]pyridine-6-carbaldehyde ≥95%
Imidazo[1,2-a]pyridine-6-carbaldehyde
Hazard: Irritant
Formula:
C₈H₆N₂O MW: 146.15 g/mol |
MDL Number:
MFCD07772808 CAS Number: 116355-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...