1-Thia-6-azaspiro[3.3]heptane 1,1-dioxide hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1,1-Dioxo-1-thia-6-azaspiro[3.3]heptane hydrochloride

115434-250MG 115434-1G
102899-310EA 2143.08 USD
102899-310 102899-312
1-Thia-6-azaspiro[3.3]heptane 1,1-dioxide hydrochloride ≥95%
1-Thia-6-azaspiro[3.3]heptane 1,1-dioxide hydrochloride

Hazard: Irritant

Formula: C₅H₉NO₂S·ClH
MW: 183.656 g/mol
MDL Number: MFCD22421741
CAS Number: 1427358-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR