Tetrahydro-1-(phenylmethyl)thiophenium hexafluorophosphate ≥95%
Supplier: Matrix Scientific
115433-1G
115433-5G
115433-25G
102899-250EA
55.38
USD
102899-250
102899-252
102899-308
Tetrahydro-1-(phenylmethyl)thiophenium hexafluorophosphate ≥95%
Tetrahydro-1-(phenylmethyl)thiophenium hexafluorophosphate
Hazard: Irritant
Formula:
C₁₁H₁₅F₆PS |
MDL Number:
MFCD22421760 CAS Number: 87301-53-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...