N-Methoxy-N,2-dimethyloxazole-4-carboxamide ≥95%

Supplier: Matrix Scientific

115432-1G 115432-5G 115432-25G
102896-178EA 140 USD
102896-178 102896-180 102896-182
N-Methoxy-N,2-dimethyloxazole-4-carboxamide ≥95%
N-Methoxy-N,2-dimethyloxazole-4-carboxamide

Hazard: Irritant

Formula: C₇H₁₀N₂O₃
MW: 170.17 g/mol
MDL Number: MFCD11040624
CAS Number: 875553-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR