4-Fluoro-2-bromophenol methyl cyclopropyl ether ≥97%
Supplier: Matrix Scientific
115431-1G
115431-5G
115431-25G
102896-172EA
70.77
USD
102896-172
102896-174
102896-176
4-Fluoro-2-bromophenol methyl cyclopropyl ether ≥97%
4-Fluoro-2-bromophenol methyl cyclopropyl ether
Hazard: Irritant
Formula:
C₁₀H₁₀BrFO |
MDL Number:
MFCD12138832 CAS Number: 1156170-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...