tert-Butyl-N-[(E)-3-tributylstannylallyl]carbamate ≥97%

Supplier: Matrix Scientific

115430-500MG 115430-1G 115430-5G
102896-166EA 316.92 USD
102896-166 102896-168 102896-170
tert-Butyl-N-[(E)-3-tributylstannylallyl]carbamate ≥97%
tert-Butyl-N-[(E)-3-tributylstannylallyl]carbamate

Hazard: Irritant

Formula: C₂₀H₄₁NO₂Sn
MW: 446.25 g/mol
MDL Number: MFCD22421664
CAS Number: 139111-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR