(1-Benzyl-3-fluoro-1,2,5,6-tetrahydropyridin-4-yl)methanol ≥95%

Supplier: Matrix Scientific

115429-500MG 115429-1G
102896-162EA 1429.23 USD
102896-162 102896-164
(1-Benzyl-3-fluoro-1,2,5,6-tetrahydropyridin-4-yl)methanol ≥95%
(1-Benzyl-3-fluoro-1,2,5,6-tetrahydropyridin-4-yl)methanol

Hazard: Irritant

Formula: C₁₃H₁₆FNO
MDL Number: MFCD20261207
CAS Number: 895578-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR