2-(2-(Trifluoromethoxy)ethyl)isoindoline-1,3-dione ≥95%

Supplier: Matrix Scientific

115427-1G 115427-5G
102896-152EA 604.62 USD
102896-152 102896-154
2-(2-(Trifluoromethoxy)ethyl)isoindoline-1,3-dione ≥95%
2-(2-(Trifluoromethoxy)ethyl)isoindoline-1,3-dione

Hazard: Irritant

Formula: C₁₁H₈F₃NO₃
MDL Number: MFCD19686683
CAS Number: 329710-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR