1-(2,2,2-Trifluoroacetyl)-3-azetidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

115426-1G 115426-5G 115426-25G
102896-146EA 70.77 USD
102896-146 102896-148 102896-150
1-(2,2,2-Trifluoroacetyl)-3-azetidinecarboxylic acid ≥95%
1-(2,2,2-Trifluoroacetyl)-3-azetidinecarboxylic acid

Hazard: Irritant

Formula: C₆H₆F₃NO₃
MDL Number: MFCD22421758
CAS Number: 1001026-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR