3-Thietanemethanamine hydrochloride ≥95%

Supplier: Matrix Scientific

115424-100MG 115424-250MG 115424-500MG
102896-136EA 769.23 USD
102896-136 102896-138 102896-140
3-Thietanemethanamine hydrochloride ≥95%
3-Thietanemethanamine hydrochloride

Hazard: Irritant

Formula: C₄H₉NS
MW: 103.19 g/mol
MDL Number: MFCD26959157
CAS Number: 1330750-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR