tert-Butyl-3-(1-hydroxy-1-methylethyl)azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(2-hydroxypropan-2-yl)azetidine-1-carboxylate, 1-Boc-3-(1-hydroxy-1-methylethyl)-azetidine

115423-1G 115423-5G
102896-132EA 1044.62 USD
102896-132 102896-134
tert-Butyl-3-(1-hydroxy-1-methylethyl)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(1-hydroxy-1-methylethyl)azetidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
MDL Number: MFCD22370133
CAS Number: 1257293-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR