2-(Oxetan-3-ylidene)acetonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
(Oxetan-3-ylidene)acetonitrile
115419-1G
115419-5G
115419-10G
102896-066EA
1567.69
USD
102896-066
102896-068
102896-070
2-(Oxetan-3-ylidene)acetonitrile ≥95%
2-(Oxetan-3-ylidene)acetonitrile
Hazard: Irritant
Formula:
C₅H₅NO MW: 95.1 g/mol |
MDL Number:
MFCD14586415 CAS Number: 1123787-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...