tert-Butyl-4-((benzyloxycarbonylamino)methyl)-3,3-difluoropiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-((((benzyloxy)carbonyl)amino)methyl)-3,3-difluoropiperidine-1-carboxylate

115415-250MG 115415-500MG 115415-1G
102896-044EA 2966.15 USD
102896-044 102896-046 102896-048
tert-Butyl-4-((benzyloxycarbonylamino)methyl)-3,3-difluoropiperidine-1-carboxylate ≥95%
tert-Butyl-4-((benzyloxycarbonylamino)methyl)-3,3-difluoropiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₉H₂₆F₂N₂O₄
MW: 384.43 g/mol
MDL Number: MFCD22199403
CAS Number: 1373503-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR