2-(1-(tert-Butoxycarbonyl)-3,3-difluoropiperidin-4-yl)acetic acid ≥95%

Supplier: Matrix Scientific

115413-500MG 115413-1G 115413-250MG
102896-034EA 3876.92 USD
102896-034 102896-036 102899-248
2-(1-(tert-Butoxycarbonyl)-3,3-difluoropiperidin-4-yl)acetic acid ≥95%
2-(1-(tert-Butoxycarbonyl)-3,3-difluoropiperidin-4-yl)acetic acid

Hazard: Corrosive

Formula: C₁₂H₁₉F₂NO₄
MDL Number: MFCD22199372
CAS Number: 1373503-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR