2-Azetidin-3-yl-propan-2-ol hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(Azetidin-3-yl)propan-2-ol hydrochloride
115412-1G
115412-5G
115412-10G
102899-374EA
989.23
USD
102899-374
102899-376
102899-378
2-Azetidin-3-yl-propan-2-ol hydrochloride ≥95%
2-Azetidin-3-yl-propan-2-ol hydrochloride
Hazard: Irritant
Formula:
C₆H₁₄ClNO MW: 151.64 g/mol |
MDL Number:
MFCD22199153 CAS Number: 1357923-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...