(1,3,3-Trimethyl-2,3-dihydro-1H-indol-5-yl)amine ≥95%
Supplier: Matrix Scientific
115411-250MG
115411-500MG
115411-1G
102899-368EA
1807.69
USD
102899-368
102899-370
102899-372
(1,3,3-Trimethyl-2,3-dihydro-1H-indol-5-yl)amine ≥95%
(1,3,3-Trimethyl-2,3-dihydro-1H-indol-5-yl)amine
Hazard: Irritant
Formula:
C₁₁H₁₆N₂ |
MDL Number:
MFCD17015862 CAS Number: 848047-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...