trans-N-Methoxy-N-methyl-2-(trifluoromethyl)cyclopropanecarboxamide ≥95%

Supplier: Matrix Scientific

115408-500MG 115408-1G 115408-5G
102895-972EA 275.38 USD
102895-972 102895-974 102895-976
trans-N-Methoxy-N-methyl-2-(trifluoromethyl)cyclopropanecarboxamide ≥95%
trans-N-Methoxy-N-methyl-2-(trifluoromethyl)cyclopropanecarboxamide

Hazard: Irritant

Formula: C₇H₁₀F₃NO₂
MW: 197.16 g/mol
MDL Number: MFCD22123286
CAS Number: 1369338-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR