(R)-1-Tetrahydrofuran-3-ylmethanamine ≥95%

Supplier: Matrix Scientific

115406-250MG 115406-500MG 115406-1G
102895-960EA 1086.15 USD
102895-960 102895-962 102895-964
(R)-1-Tetrahydrofuran-3-ylmethanamine ≥95%
(R)-1-Tetrahydrofuran-3-ylmethanamine

Hazard: Irritant

Formula: C₅H₁₁NO
MDL Number: MFCD17015963
CAS Number: 1048962-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR