1H-Pyrrolo[2,3-b]pyridine-2-carboxaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 1H-Pyrrolo[2,3-b]pyridine-2-carbaldehyde

115403-1G 115403-5G
102896-238EA 989.23 USD
102896-238 102896-240
1H-Pyrrolo[2,3-b]pyridine-2-carboxaldehyde ≥95%
1H-Pyrrolo[2,3-b]pyridine-2-carboxaldehyde

Hazard: Irritant

Formula: C₈H₆N₂O
MW: 146.15 g/mol
MDL Number: MFCD08437634
CAS Number: 394223-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR