4-(Oxetan-3-yl)piperazine ditrifluoroacetic acid salt ≥95%
Supplier: Matrix Scientific
115401-100MG
115401-250MG
115401-500MG
102896-228EA
769.23
USD
102896-228
102896-230
102896-232
4-(Oxetan-3-yl)piperazine ditrifluoroacetic acid salt ≥95%
4-(Oxetan-3-yl)piperazine ditrifluoroacetic acid salt
Hazard: Irritant
Formula:
C₁₁H₁₆F₆N₂O₅ MW: 370.25 g/mol |
MDL Number:
MFCD22573363 CAS Number: 1404373-75-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...