3,3-Oxetanedimethanamine dihydrochloride ≥95%
Supplier: Matrix Scientific
115400-250MG
115400-500MG
115400-1G
102896-222EA
769.23
USD
102896-222
102896-224
102896-226
3,3-Oxetanedimethanamine dihydrochloride ≥95%
3,3-Oxetanedimethanamine dihydrochloride
Hazard: Irritant
Formula:
C₅H₁₄Cl₂N₂O MW: 189.09 g/mol |
MDL Number:
MFCD18643362 CAS Number: 111511-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...