Tetrahydropyran-4-ylmethyl methanesulfonate ≥95%

Supplier: Matrix Scientific

115399-500MG 115399-1G 115399-5G
102896-216EA 275.38 USD
102896-216 102896-218 102896-220
Tetrahydropyran-4-ylmethyl methanesulfonate ≥95%
Tetrahydropyran-4-ylmethyl methanesulfonate

Hazard: Irritant

Formula: C₇H₁₄O₄S
MDL Number: MFCD18785628
CAS Number: 132291-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR