(3aR,7aR)-Octahydro-3H-pyrrolo[3,4-c]pyridin-3-one ≥95%

Supplier: Matrix Scientific

115398-500MG 115398-1G
102896-212EA 966.88 USD
102896-212 102896-214
(3aR,7aR)-Octahydro-3H-pyrrolo[3,4-c]pyridin-3-one ≥95%
(3aR,7aR)-Octahydro-3H-pyrrolo[3,4-c]pyridin-3-one

Hazard: Irritant

Formula: C₇H₁₂N₂O
MDL Number: MFCD13178722
CAS Number: 868552-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR