3-Methyl-3-oxetanamine hemioxalate ≥97%

Supplier: Matrix Scientific

115396-500MG 115396-1G 115396-250MG
102895-944EA 724.62 USD
102895-944 102895-946 102899-362
3-Methyl-3-oxetanamine hemioxalate ≥97%
3-Methyl-3-oxetanamine hemioxalate

Hazard: Irritant

Formula: C₆H₁₁NO₅
MDL Number: MFCD22422283
CAS Number: 1369338-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR