Methyl-6-bromopyrazolo[1,5-a]pyridine-3-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 6-bromopyrazolo[1,5-a]pyridine-3-carboxylate
115394-500MG
115394-1G
102899-350EA
1238.46
USD
102899-350
102899-352
Methyl-6-bromopyrazolo[1,5-a]pyridine-3-carboxylate ≥95%
Methyl-6-bromopyrazolo[1,5-a]pyridine-3-carboxylate
Hazard: Irritant
Formula:
C₉H₇BrN₂O₂ MW: 255.08 g/mol |
MDL Number:
MFCD15474972 CAS Number: 1062368-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...