3-(Hydroxymethyl)-3-methylpiperidine ≥95%
Supplier: Matrix Scientific
115391-500MG
115391-1G
115391-5G
102899-300EA
824.62
USD
102899-300
102899-302
102899-304
3-(Hydroxymethyl)-3-methylpiperidine ≥95%
3-(Hydroxymethyl)-3-methylpiperidine
Hazard: Irritant
Formula:
C₇H₁₅NO |
MDL Number:
MFCD09055357 CAS Number: 221298-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...