3-Formylindole-5-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-formyl-1H-indole-5-carboxylate

115390-1G 115390-5G 115390-10G
102895-940EA 798.46 USD
102895-940 102895-942 102899-298
3-Formylindole-5-carboxylic acid methyl ester ≥95%
3-Formylindole-5-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₁₁H₉NO₃
MW: 203.2 g/mol
MDL Number: MFCD00211075
CAS Number: 197506-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR