Ethyl-5-bromopyrazolo[1,5-a]pyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-bromopyrazolo[1,5-a]pyridine-3-carboxylate

115388-250MG 115388-500MG 115388-1G
102895-928EA 715.38 USD
102895-928 102895-930 102895-932
Ethyl-5-bromopyrazolo[1,5-a]pyridine-3-carboxylate ≥95%
Ethyl-5-bromopyrazolo[1,5-a]pyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₉BrN₂O₂
MW: 269.1 g/mol
MDL Number: MFCD06739182
CAS Number: 885276-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR