Methyl-4-bromopyrazolo[1,5-a]pyridine-3-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-bromopyrazolo[1,5-a]pyridine-3-carboxylate
115387-250MG
115387-500MG
115387-1G
102895-922EA
1318.46
USD
102895-922
102895-924
102895-926
Methyl-4-bromopyrazolo[1,5-a]pyridine-3-carboxylate ≥95%
Methyl-4-bromopyrazolo[1,5-a]pyridine-3-carboxylate
Hazard: Irritant
Formula:
C₉H₇BrN₂O₂ MW: 255.08 g/mol |
MDL Number:
MFCD19703856 CAS Number: 1062368-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...