Diphenylvinylsulfonium triflate 90-95%
Supplier: Matrix Scientific
Synonyms:
Diphenyl(vinyl)sulphonium trifluoromethanesulphonate
115386-500MG
115386-1G
115386-5G
102895-868EA
387.69
USD
102895-868
102895-918
102895-920
Diphenylvinylsulfonium triflate 90-95%
Diphenylvinylsulfonium triflate
Hazard: Irritant
Formula:
C₁₅H₁₃F₃O₃S₂ MW: 362.39 g/mol |
MDL Number:
MFCD22123284 CAS Number: 247129-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...