3,3-Difluoro-5-hydroxypiperidin-2-one ≥95%

Supplier: Matrix Scientific

115384-500MG 115384-1G
102895-860EA 4393.85 USD
102895-860 102895-862
3,3-Difluoro-5-hydroxypiperidin-2-one ≥95%
3,3-Difluoro-5-hydroxypiperidin-2-one

Hazard: Irritant

Formula: C₅H₇F₂NO₂
MW: 151.11 g/mol
MDL Number: MFCD18433741
CAS Number: 1256080-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR