3,3-Difluoro-1-boc-azetidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3,3-difluoroazetidine-1-carboxylate

115383-1G 115383-5G 115383-10G
102895-854EA 304.62 USD
102895-854 102895-856 102895-858
3,3-Difluoro-1-boc-azetidine ≥95%
3,3-Difluoro-1-boc-azetidine

Hazard: Irritant

Formula: C₈H₁₃F₂NO₂
MW: 193.2 g/mol
MDL Number: MFCD18381087
CAS Number: 1255666-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR