3-Chloro-5-(chloromethyl)pyridine ≥95%

Supplier: Matrix Scientific

115380-1G 115380-5G
102896-206EA 1300 USD
102896-206 102896-208
3-Chloro-5-(chloromethyl)pyridine ≥95%
3-Chloro-5-(chloromethyl)pyridine

Hazard: Irritant

Formula: C₆H₅Cl₂N
MDL Number: MFCD10697605
CAS Number: 847737-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR