tert-Butyl-3-(2-oxopyrrolidin-1-yl)azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115379-250MG 115379-500MG
102896-200EA 1292.31 USD
102896-200 102896-202
tert-Butyl-3-(2-oxopyrrolidin-1-yl)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(2-oxopyrrolidin-1-yl)azetidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₀N₂O₃
MW: 240.3 g/mol
MDL Number: MFCD22123279
CAS Number: 1257294-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR