tert-Butyl-3-(4-hydroxy-1-piperidyl)azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115378-100MG 115378-250MG 115378-500MG
102896-120EA 580.35 USD
102896-120 102896-122 102896-124
tert-Butyl-3-(4-hydroxy-1-piperidyl)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(4-hydroxy-1-piperidyl)azetidine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₃
MW: 256.35 g/mol
MDL Number: MFCD09264405
CAS Number: 178311-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR