tert-Butyl-3-(4-hydroxy-4-methyl-1-piperidyl)azetidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
115377-100MG
115377-250MG
115377-500MG
102896-114EA
824.62
USD
102896-114
102896-116
102896-118
tert-Butyl-3-(4-hydroxy-4-methyl-1-piperidyl)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(4-hydroxy-4-methyl-1-piperidyl)azetidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₄H₂₆N₂O₃ |
MDL Number:
MFCD13194997 CAS Number: 550371-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...