tert-Butyl-3-(3-hydroxy-3-methylazetidin-1-yl)azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115376-100MG 115376-250MG 115376-500MG
102896-108EA 824.62 USD
102896-108 102896-110 102896-112
tert-Butyl-3-(3-hydroxy-3-methylazetidin-1-yl)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(3-hydroxy-3-methylazetidin-1-yl)azetidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₃
MDL Number: MFCD22123281
CAS Number: 1257294-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR