tert-Butyl-3-(3-hydroxyazetidin-1-yl)azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115375-100MG 115375-250MG 115375-500MG
102895-844EA 824.62 USD
102895-844 102896-104 102896-106
tert-Butyl-3-(3-hydroxyazetidin-1-yl)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(3-hydroxyazetidin-1-yl)azetidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₃
MW: 228.29 g/mol
MDL Number: MFCD22123275
CAS Number: 1257293-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR