tert-Butyl-3-(4-fluoro-1-piperidyl)azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115374-250MG 115374-500MG 115374-1G
102895-838EA 344.62 USD
102895-838 102895-840 102895-842
tert-Butyl-3-(4-fluoro-1-piperidyl)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(4-fluoro-1-piperidyl)azetidine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₃FN₂O₂
MW: 258.34 g/mol
MDL Number: MFCD22123270
CAS Number: 1257293-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR