tert-Butyl-3-(3-fluoroazetidin-1-yl)azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115373-100MG 115373-250MG 115373-500MG
102895-832EA 824.62 USD
102895-832 102895-834 102895-836
tert-Butyl-3-(3-fluoroazetidin-1-yl)azetidine-1-carboxylate ≥95%
tert-Butyl-3-(3-fluoroazetidin-1-yl)azetidine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₉FN₂O₂
MW: 230.28 g/mol
MDL Number: MFCD22123269
CAS Number: 1257293-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR